Assay Details
| Gene Symbol: Tbx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285472.1 | NP_001272401.1 | 6 - 7 | 930 | 66 | |
| NM_001285476.1 | NP_001272405.1 | 4 - 5 | 854 | 66 | ||
| NM_011532.2 | NP_035662.1 | 4 - 5 | 858 | 66 | ||
| GenBank mRNA | AF326960.2 | - | 4 - 5 | 858 | 66 | |
| AF349658.1 | - | 6 - 7 | 930 | 66 | ||
| BC139474.1 | - | 4 - 5 | 858 | 8862496 | 66 | |
| BC145636.1 | - | 4 - 5 | 854 | 66 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Tbx1
Genomic Map