Assay Details
| Gene Symbol: Yars2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198246.2 | NP_937889.1 | 4 - 5 | 1278 | 75 | |
| NR_038046.1 | 4 - 5 | 1243 | 75 | |||
| GenBank mRNA | AK039123.1 | - | 4 - 5 | 1053 | 75 | |
| AK138504.1 | - | 4 - 5 | 1228 | 75 | ||
| AK210912.1 | - | 1 - 2 | 67 | 75 | ||
| BC060295.1 | - | 4 - 5 | 1261 | 30290485 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
translation
tRNA aminoacylation for protein translation
tyrosyl-tRNA aminoacylation
tRNA aminoacylation
mitochondrial tyrosyl-tRNA aminoacylation
tRNA binding
nucleotide binding
RNA binding
aminoacyl-tRNA ligase activity
tyrosine-tRNA ligase activity
ATP binding
ligase activity
protein homodimerization activity
poly(A) RNA binding
tyrosine binding
translation
tRNA aminoacylation for protein translation
tyrosyl-tRNA aminoacylation
tRNA aminoacylation
mitochondrial tyrosyl-tRNA aminoacylation
tRNA binding
nucleotide binding
RNA binding
aminoacyl-tRNA ligase activity
tyrosine-tRNA ligase activity
ATP binding
ligase activity
protein homodimerization activity
poly(A) RNA binding
tyrosine binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30568848 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30568847 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30568846 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30568845 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30568844 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30568843 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P373609 | BC060295.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P373609 | AK083653.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P373609 | AK138504.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P373609 | AK212968.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P373609 | NM_198246.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P373609 | BC060295.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P373609 | AK083653.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P373609 | AK138504.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P373609 | AK212968.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P373609 | NM_198246.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 765995 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 130460 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 16493 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 170647 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 728338 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P124260 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P373609 | BC060295.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P373609 | AK083653.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P373609 | AK138504.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P373609 | AK212968.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P373609 | NM_198246.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P373609 | BC060295.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P373609 | AK083653.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P373609 | AK138504.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P373609 | AK212968.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P373609 | NM_198246.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P373609 | BC060295.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P373609 | AK083653.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P373609 | AK138504.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P373609 | AK212968.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P373609 | NM_198246.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P373609 | BC060295.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P373609 | AK083653.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P373609 | AK138504.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P373609 | AK212968.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P373609 | NM_198246.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30568857 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30568839 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30568829 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30568834 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30568845 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30568861 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30568841 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30568848 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30568858 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30568852 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30568855 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30568847 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30568862 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30568836 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30568840 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30568838 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30568830 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30568844 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30568832 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30568850 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08518406 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P124260 | - | - |
| Whole Mouse Genome G4122A | A_51_P373609 | BC060295.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P373609 | AK083653.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P373609 | AK138504.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P373609 | AK212968.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P373609 | NM_198246.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P373609 | BC060295.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P373609 | AK083653.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P373609 | AK138504.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P373609 | AK212968.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P373609 | NM_198246.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P913 | AK083653.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P111698 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P124260 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13009150 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13009154 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12962986 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12962984 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12962975 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30568848 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30568846 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30568843 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30568844 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30568847 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30568845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30568888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30568850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30568885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30568859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30568879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30568852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30568882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30568845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30568834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30568846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30568837 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30568832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30568848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30568878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30568861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30568883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30568876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30568847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30568867 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30568841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30568874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30568857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30568877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30568855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30568862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30568829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30568839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30568819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30568890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30568871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30568853 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12962979 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12639576 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30568867 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12659114 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30568859 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30568877 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30568874 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30568832 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30568886 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30568852 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30568879 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30568863 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30568864 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12659398 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30568876 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30568850 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30568819 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30568834 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30568866 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30568858 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12962984 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30568823 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30568826 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30568857 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30568855 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30568829 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12659409 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30568882 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30568890 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13009154 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12962980 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30568837 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30568891 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30568861 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30568883 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30568885 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30568862 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30568888 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30568821 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12962977 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12855135 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30568872 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10433908 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10433909 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10433905 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10433906 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10433907 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1439442_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1430134_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10433904 | - | - |
| Mouse Genome 430 2.0 Array | 1439442_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1439442_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1430134_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1430134_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 70120_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 70120_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1600001525.mm.1 | NM_198246.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600001525.mm.1 | NM_198246.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600001525.mm.1 | NR_038046.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600001525.mm.1 | NR_038046.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600001525.mm.1 | TC1600000196.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1430134_a_at | - | - |
| Mouse Expression Set 430 | 1430134_PM_a_at | - | - |
| Mouse Expression Set 430 | 1439442_x_at | - | - |
| Mouse Expression Set 430 | 1439442_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430134_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1439442_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430134_a_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1439442_x_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 527170 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 102640142 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 102640142 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6100731 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006100731 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2712064 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1244637 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2712064 | - | - |
| mouse-6 v1.1 (Illumina) | scl49389.6.1_58-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 102640142 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6100731 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl49389.6.1_58-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 102640142 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6100731 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2712064 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1244637 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2712064 | - | - |

Yars2
Genomic Map