Assay Details
| Gene Symbol: Fgd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF402611.1 | - | 9 - 10 | 1518 | 88 | |
| AF402612.1 | - | 8 - 9 | 1415 | 88 | ||
| AF402613.1 | - | 9 - 10 | 1565 | 88 | ||
| AK028302.1 | - | 6 - 7 | 967 | 88 | ||
| AK034504.1 | - | 8 - 9 | 1508 | 88 | ||
| AK134961.1 | - | 9 - 10 | 1581 | 88 | ||
| AK149626.1 | - | 9 - 10 | 1569 | 88 | ||
| BC129902.1 | - | 9 - 10 | 1605 | 40111637 | 88 | |
| BC129903.1 | - | 9 - 10 | 1606 | 40111639 | 88 | |
| BC145523.1 | - | 9 - 10 | 1659 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fgd4
Genomic Map