Assay Details
| Gene Symbol: Dnm1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025947.2 | NP_001021118.1 | 6 - 7 | 787 | 86 | |
| NM_001276340.1 | NP_001263269.1 | 6 - 7 | 805 | 86 | ||
| NM_001276341.1 | NP_001263270.1 | 5 - 6 | 549 | 86 | ||
| NM_152816.3 | NP_690029.2 | 7 - 8 | 826 | 86 | ||
| NR_075074.1 | 6 - 7 | 596 | 86 | |||
| GenBank mRNA | AB079133.1 | - | 6 - 7 | 639 | 86 | |
| AK018195.1 | - | 7 - 8 | 86 | |||
| AK051443.1 | - | 6 - 7 | 716 | 86 | ||
| AK080871.1 | - | 6 - 7 | 707 | 86 | ||
| AK154123.1 | - | 6 - 7 | 556 | 86 | ||
| BC040777.1 | - | 5 - 6 | 519 | 4948368 | 86 | |
| BC079635.1 | - | 6 - 7 | 636 | 6400313 | 86 | |
| BC085158.1 | - | 7 - 8 | 731 | 86 | ||
| CK634757.1 | - | 4 - 5 | 358 | 86 | ||
| CN525473.1 | - | 1 - 2 | 177 | 86 | ||
| CO432650.1 | - | 3 - 4 | 315 | 86 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Dnm1l
Genomic Map