Assay Details
| Gene Symbol: Rnf20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163263.1 | NP_001156735.1 | 10 - 11 | 1753 | 61 | |
| NM_182999.2 | NP_892044.1 | 11 - 12 | 1600 | 61 | ||
| GenBank mRNA | AK048862.1 | - | 10 - 11 | 1712 | 61 | |
| AK076501.1 | - | 6 - 7 | 682 | 61 | ||
| AK139883.1 | - | 8 - 9 | 1128 | 61 | ||
| AK163905.1 | - | 7 - 8 | 965 | 61 | ||
| AK220492.1 | - | 10 - 11 | 1669 | 61 | ||
| BC004593.1 | - | 6 - 7 | 827 | 61 | ||
| BC052482.1 | - | 11 - 12 | 1539 | 6308280 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rnf20
Genomic Map