Assay Details
| Gene Symbol: Aldob | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144903.2 | NP_659152.1 | 3 - 4 | 385 | 103 | |
| GenBank mRNA | AK135385.1 | - | 3 - 4 | 414 | 103 | |
| AK149395.1 | - | 3 - 4 | 402 | 103 | ||
| AK149598.1 | - | 3 - 4 | 399 | 103 | ||
| AK167334.1 | - | 3 - 4 | 402 | 103 | ||
| AK167566.1 | - | 3 - 4 | 399 | 103 | ||
| AK168600.1 | - | 3 - 4 | 399 | 103 | ||
| AK168997.1 | - | 3 - 4 | 449 | 103 | ||
| BC016435.1 | - | 3 - 4 | 381 | 4241025 | 103 | |
| BC022113.1 | - | 3 - 4 | 358 | 5098524 | 103 | |
| BC024056.1 | - | 3 - 4 | 391 | 5100067 | 103 | |
| BC024112.1 | - | 3 - 4 | 390 | 5102118 | 103 | |
| BC026577.1 | - | 3 - 4 | 350 | 4989521 | 103 | |
| BC030724.1 | - | 3 - 4 | 387 | 5100784 | 103 | |
| BC030725.1 | - | 3 - 4 | 391 | 5101686 | 103 | |
| BC034169.1 | - | 3 - 4 | 391 | 4975382 | 103 | |
| BC034171.1 | - | 3 - 4 | 368 | 4972521 | 103 | |
| BC034172.1 | - | 3 - 4 | 355 | 4974489 | 103 | |
| BC034173.1 | - | 3 - 4 | 367 | 4974553 | 103 | |
| BC036130.1 | - | 3 - 4 | 368 | 4974071 | 103 | |
| BC036131.1 | - | 3 - 4 | 385 | 4974877 | 103 | |
| BC036132.1 | - | 3 - 4 | 385 | 4975336 | 103 | |
| BC036133.1 | - | 3 - 4 | 397 | 4975573 | 103 | |
More Information
Updated Mapping Information
Set Membership: |
|

Aldob
Genomic Map