Assay Details
| Gene Symbol: Aldob | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144903.2 | NP_659152.1 | 7 - 8 | 857 | 68 | |
| GenBank mRNA | AK135385.1 | - | 7 - 8 | 886 | 68 | |
| AK149395.1 | - | 7 - 8 | 874 | 68 | ||
| AK149598.1 | - | 7 - 8 | 871 | 68 | ||
| AK167334.1 | - | 7 - 8 | 874 | 68 | ||
| AK167566.1 | - | 7 - 8 | 871 | 68 | ||
| AK168600.1 | - | 7 - 8 | 871 | 68 | ||
| AK168997.1 | - | 7 - 8 | 921 | 68 | ||
| BC016435.1 | - | 7 - 8 | 853 | 4241025 | 68 | |
| BC022113.1 | - | 7 - 8 | 830 | 5098524 | 68 | |
| BC024056.1 | - | 7 - 8 | 863 | 5100067 | 68 | |
| BC024112.1 | - | 7 - 8 | 862 | 5102118 | 68 | |
| BC026577.1 | - | 7 - 8 | 822 | 4989521 | 68 | |
| BC030724.1 | - | 7 - 8 | 859 | 5100784 | 68 | |
| BC030725.1 | - | 7 - 8 | 863 | 5101686 | 68 | |
| BC034169.1 | - | 7 - 8 | 863 | 4975382 | 68 | |
| BC034171.1 | - | 7 - 8 | 840 | 4972521 | 68 | |
| BC034172.1 | - | 7 - 8 | 827 | 4974489 | 68 | |
| BC034173.1 | - | 7 - 8 | 839 | 4974553 | 68 | |
| BC036130.1 | - | 7 - 8 | 840 | 4974071 | 68 | |
| BC036131.1 | - | 7 - 8 | 857 | 4974877 | 68 | |
| BC036132.1 | - | 7 - 8 | 857 | 4975336 | 68 | |
| BC036133.1 | - | 7 - 8 | 869 | 4975573 | 68 | |
| BC036139.1 | - | 3 - 4 | 481 | 68 | ||
| BC036142.1 | - | 1 - 2 | 164 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Aldob
Genomic Map