Assay Details
| Gene Symbol: Pvrl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021495.4 | NP_067470.1 | 3 - 4 | 994 | 128 | |
| NM_021496.3 | NP_067471.1 | 3 - 4 | 994 | 128 | ||
| NM_021497.2 | NP_067472.1 | 3 - 4 | 994 | 128 | ||
| GenBank mRNA | AF195833.1 | - | 3 - 4 | 991 | 128 | |
| AF195834.1 | - | 3 - 4 | 801 | 128 | ||
| AF195835.1 | - | 3 - 4 | 801 | 128 | ||
| AK011949.1 | - | 3 - 4 | 994 | 128 | ||
| BC125588.1 | - | 3 - 4 | 920 | 40130103 | 128 | |
| BC132187.1 | - | 3 - 4 | 920 | 40130464 | 128 | |
| BU701648.1 | - | 2 - 3 | 360 | 128 | ||
More Information
Set Membership: |
|

Pvrl3
Genomic Map