Assay Details
| Gene Symbol: Muc13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010739.2 | NP_034869.1 | 6 - 7 | 1184 | 123 | |
| GenBank mRNA | AK132274.1 | - | 5 - 6 | 1029 | 123 | |
| BC024321.1 | - | 5 - 6 | 1150 | 4219376 | 123 | |
| J04634.1 | - | 6 - 7 | 1184 | 123 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Muc13
Genomic Map