| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1600012607.mm.1 |
NM_001164268.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600012607.mm.1 |
NM_001164268.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600012607.mm.1 |
NM_177357.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600012607.mm.1 |
NM_177357.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600012607.mm.1 |
TC1600001528.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600012615.mm.1 |
NM_001164268.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600012615.mm.1 |
NM_001164268.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600012615.mm.1 |
NM_177357.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600012615.mm.1 |
NM_177357.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600012615.mm.1 |
TC1600001528.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600012608.mm.1 |
NM_001164268.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600012608.mm.1 |
NM_001164268.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600012608.mm.1 |
NM_177357.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600012608.mm.1 |
NM_177357.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600012608.mm.1 |
TC1600001528.mm.1 |
MTA TC |