Assay Details
| Gene Symbol: Heg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175256.5 | NP_780465.4 | 13 - 14 | 3071 | 125 | |
| GenBank mRNA | AK043598.1 | - | 2 - 3 | 1309 | 125 | |
| BC072651.1 | - | 5 - 6 | 381 | 125 | ||
| BC076596.1 | - | 11 - 12 | 2350 | 30645937 | 125 | |
More Information
Updated Mapping Information
Set Membership: |
|

Heg1
Genomic Map