Assay Details
| Gene Symbol: Heg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175256.5 | NP_780465.4 | 3 - 4 | 910 | 93 | |
| GenBank mRNA | BC076596.1 | - | 1 - 2 | 189 | 30645937 | 93 |
| BE862445.1 | - | 1 - 2 | 93 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC076596.1 transcripts.
Set Membership: |
|

Heg1
Genomic Map