Assay Details
| Gene Symbol: Heg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175256.5 | NP_780465.4 | 5 - 6 | 2322 | 127 | |
| GenBank mRNA | BC076596.1 | - | 3 - 4 | 1601 | 30645937 | 127 |
More Information
Updated Mapping Information
Set Membership: |
|

Heg1
Genomic Map