Assay Details
| Gene Symbol: Cblb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033238.1 | NP_001028410.1 | 8 - 9 | 1217 | 103 | |
| GenBank mRNA | AK147367.1 | - | 8 - 9 | 1203 | 103 | |
| AK156295.1 | - | 9 - 10 | 1248 | 103 | ||
| AK161486.1 | - | 6 - 7 | 803 | 103 | ||
| AK170640.1 | - | 8 - 9 | 1161 | 103 | ||
| AK171350.1 | - | 8 - 9 | 1162 | 103 | ||
| BC150934.1 | - | 8 - 9 | 1173 | 9087845 | 103 | |
| BC150938.1 | - | 8 - 9 | 1173 | 9087849 | 103 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cblb
Genomic Map