Assay Details
| Gene Symbol: Mast3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199308.2 | NP_955012.2 | 10 - 11 | 1188 | 96 | |
| GenBank mRNA | AK140821.1 | - | 10 - 11 | 1197 | 96 | |
| AK155568.1 | - | 11 - 12 | 1160 | 96 | ||
More Information
Set Membership: |
|

Mast3
Genomic Map