Assay Details
| Gene Symbol: Mast3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199308.2 | NP_955012.2 | 11 - 12 | 1275 | 71 | |
| GenBank mRNA | AK140821.1 | - | 11 - 12 | 1284 | 71 | |
| AK155568.1 | - | 12 - 13 | 1247 | 71 | ||
| CB249116.1 | - | 1 - 2 | 80 | 71 | ||
More Information
Set Membership: |
|

Mast3
Genomic Map