Assay Details
| Gene Symbol: Mast3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199308.2 | NP_955012.2 | 13 - 14 | 1619 | 55 | |
| GenBank mRNA | AK140821.1 | - | 13 - 14 | 1628 | 55 | |
| AK155568.1 | - | 14 - 15 | 1591 | 55 | ||
| CB249116.1 | - | 3 - 4 | 424 | 55 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mast3
Genomic Map