Assay Details
| Gene Symbol: Mast3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199308.2 | NP_955012.2 | 15 - 16 | 1919 | 59 | |
| GenBank mRNA | AK140821.1 | - | 15 - 16 | 1928 | 59 | |
| AK155568.1 | - | 16 - 17 | 1891 | 59 | ||
More Information
Set Membership: |
|

Mast3
Genomic Map