Assay Details
| Gene Symbol: Mast3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199308.2 | NP_955012.2 | 22 - 23 | 3068 | 102 | |
| GenBank mRNA | AK140821.1 | - | 22 - 23 | 3082 | 102 | |
| AK172971.1 | - | 2 - 3 | 280 | 102 | ||
| BC024265.1 | - | 4 - 5 | 716 | 102 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC024265.1 transcripts.
Set Membership: |
|

Mast3
Genomic Map