Assay Details
| Gene Symbol: Adcy5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012765.4 | NP_001012783.3 | 14 - 15 | 3189 | 54 | |
| GenBank mRNA | AK147590.1 | - | 3292 | 54 | ||
| AK147649.1 | - | 3949 | 54 | |||
| AK160942.1 | - | 14 - 15 | 3189 | 54 | ||
| BC035550.1 | - | 1327 | 54 | |||
| BC090846.1 | - | 14 - 15 | 2924 | 30363147 | 54 | |
More Information
Updated Mapping Information
Set Membership: |
|

Adcy5
Genomic Map