Assay Details
| Gene Symbol: Sec22a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133704.4 | NP_598465.2 | 5 - 6 | 774 | 90 | |
| GenBank mRNA | AK044615.1 | - | 5 - 6 | 709 | 90 | |
| AK082516.1 | - | 5 - 6 | 774 | 90 | ||
| BC010189.1 | - | 5 - 6 | 503 | 90 | ||
| BC138969.1 | - | 4 - 5 | 688 | 8861991 | 90 | |
| BC138971.1 | - | 4 - 5 | 688 | 8861993 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sec22a
Genomic Map