Assay Details
| Gene Symbol: St3gal6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018784.2 | NP_061254.1 | 7 - 8 | 930 | 113 | |
| GenBank mRNA | AB063326.1 | - | 5 - 6 | 433 | 113 | |
| AF119390.1 | - | 7 - 8 | 930 | 113 | ||
| AK033562.1 | - | 7 - 8 | 817 | 113 | ||
| AK041173.1 | - | 7 - 8 | 973 | 113 | ||
| AK048950.1 | - | 7 - 8 | 974 | 113 | ||
| AK162619.1 | - | 8 - 9 | 910 | 113 | ||
| AK197012.1 | - | 3 - 4 | 206 | 113 | ||
| BC052338.1 | - | 6 - 7 | 691 | 30061291 | 113 | |
More Information
Updated Mapping Information
Set Membership: |
|

St3gal6
Genomic Map