Assay Details
| Gene Symbol: Pros1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011173.2 | NP_035303.1 | 1 - 2 | 172 | 78 | |
| GenBank mRNA | AK145501.1 | - | 1 - 2 | 172 | 78 | |
| AK163032.1 | - | 1 - 2 | 200 | 78 | ||
| BC128316.1 | - | 1 - 2 | 134 | 40109789 | 78 | |
| Z25469.1 | - | 1 - 2 | 138 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pros1
Genomic Map