Assay Details
| Gene Symbol: Pros1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011173.2 | NP_035303.1 | 3 - 4 | 352 | 92 | |
| GenBank mRNA | AK145501.1 | - | 3 - 4 | 352 | 92 | |
| AK163032.1 | - | 3 - 4 | 380 | 92 | ||
| BC128316.1 | - | 3 - 4 | 314 | 40109789 | 92 | |
| L27439.1 | - | 2 - 3 | 163 | 92 | ||
| Z25469.1 | - | 3 - 4 | 318 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pros1
Genomic Map