Assay Details
| Gene Symbol: Pros1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011173.2 | NP_035303.1 | 4 - 5 | 436 | 69 | |
| GenBank mRNA | AK145501.1 | - | 4 - 5 | 436 | 69 | |
| AK163032.1 | - | 4 - 5 | 464 | 69 | ||
| BC128316.1 | - | 4 - 5 | 398 | 40109789 | 69 | |
| L27439.1 | - | 3 - 4 | 247 | 69 | ||
| Z25469.1 | - | 4 - 5 | 402 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pros1
Genomic Map