Assay Details
| Gene Symbol: Slc15a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021301.3 | NP_067276.2 | 9 - 10 | 1077 | 68 | |
| GenBank mRNA | AF111811.1 | - | 9 - 10 | 1077 | 68 | |
| AK128939.1 | - | 9 - 10 | 910 | 68 | ||
| BC138627.1 | - | 9 - 10 | 1034 | 8861647 | 68 | |
| BC138628.1 | - | 9 - 10 | 1034 | 8861648 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc15a2
Genomic Map