Assay Details
| Gene Symbol: Rabl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042499.1 | NP_001035964.1 | 4 - 5 | 404 | 87 | |
| GenBank mRNA | AK016099.1 | - | 4 - 5 | 413 | 87 | |
| AK028195.1 | - | 4 - 5 | 398 | 87 | ||
| AK028269.1 | - | 4 - 5 | 391 | 87 | ||
| BC050194.1 | - | 4 - 5 | 385 | 30022531 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rabl3
Genomic Map