Assay Details
| Gene Symbol: Rabl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042499.1 | NP_001035964.1 | 5 - 6 | 557 | 130 | |
| GenBank mRNA | AK011280.1 | - | 5 - 6 | 499 | 130 | |
| AK016099.1 | - | 5 - 6 | 566 | 130 | ||
| AK028195.1 | - | 5 - 6 | 551 | 130 | ||
| AK028269.1 | - | 5 - 6 | 544 | 130 | ||
| BC050194.1 | - | 5 - 6 | 538 | 30022531 | 130 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rabl3
Genomic Map