Assay Details
| Gene Symbol: Hcls1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008225.2 | NP_032251.2 | 2 - 3 | 176 | 66 | |
| GenBank mRNA | AK040049.1 | - | 2 - 3 | 164 | 66 | |
| AK088723.1 | - | 2 - 3 | 158 | 66 | ||
| BC007469.1 | - | 2 - 3 | 146 | 3593456 | 66 | |
| D42120.1 | - | 2 - 3 | 107 | 66 | ||
| X84797.1 | - | 2 - 3 | 144 | 66 | ||
More Information
Set Membership: |
|

Hcls1
Genomic Map