Assay Details
| Gene Symbol: Hcls1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008225.2 | NP_032251.2 | 3 - 4 | 246 | 113 | |
| GenBank mRNA | AK040049.1 | - | 3 - 4 | 234 | 113 | |
| AK088723.1 | - | 3 - 4 | 228 | 113 | ||
| BC007469.1 | - | 3 - 4 | 216 | 3593456 | 113 | |
| D42120.1 | - | 3 - 4 | 177 | 113 | ||
| X84797.1 | - | 3 - 4 | 214 | 113 | ||
More Information
Set Membership: |
|

Hcls1
Genomic Map