Assay Details
| Gene Symbol: Masp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008555.2 | NP_032581.2 | 7 - 8 | 1067 | 87 | |
| GenBank mRNA | AK031598.1 | - | 7 - 8 | 1128 | 87 | |
| AK043055.1 | - | 7 - 8 | 1127 | 87 | ||
| BC131637.1 | - | 7 - 8 | 1067 | 40109637 | 87 | |
| BC131638.1 | - | 7 - 8 | 1067 | 40109639 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Masp1
Genomic Map