Assay Details
| Gene Symbol: Masp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008555.2 | NP_032581.2 | 8 - 9 | 1146 | 87 | |
| GenBank mRNA | AB049755.1 | - | 8 - 9 | 1159 | 87 | |
| AK031598.1 | - | 8 - 9 | 1207 | 87 | ||
| BC131637.1 | - | 8 - 9 | 1146 | 40109637 | 87 | |
| BC131638.1 | - | 8 - 9 | 1146 | 40109639 | 87 | |
| D16492.1 | - | 1159 | 87 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Masp1
Genomic Map