Assay Details
| Gene Symbol: Adipoq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009605.4 | NP_033735.3 | 1 - 2 | 110 | 71 | |
| GenBank mRNA | AK041214.1 | - | 1 - 2 | 60 | 71 | |
| AK134112.1 | - | 1 - 2 | 110 | 71 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK041214.1, AK134112.1, NM_009605.4 transcripts.
Set Membership: |
|

Adipoq
Genomic Map