Assay Details
| Gene Symbol: Dvl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007889.2 | NP_031915.2 | 4 - 5 | 595 | 126 | |
| GenBank mRNA | AK052151.1 | - | 4 - 5 | 596 | 126 | |
| BC033414.1 | - | 3 - 4 | 1557 | 126 | ||
| U41285.1 | - | 4 - 5 | 677 | 126 | ||
More Information
Set Membership: |
|

Dvl3
Genomic Map