Assay Details
| Gene Symbol: Dvl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007889.2 | NP_031915.2 | 7 - 8 | 889 | 87 | |
| GenBank mRNA | AK052151.1 | - | 7 - 8 | 890 | 87 | |
| U41285.1 | - | 7 - 8 | 971 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dvl3
Genomic Map