Assay Details
| Gene Symbol: Dvl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007889.2 | NP_031915.2 | 8 - 9 | 1033 | 65 | |
| GenBank mRNA | AK052151.1 | - | 8 - 9 | 1034 | 65 | |
| U41285.1 | - | 8 - 9 | 1115 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dvl3
Genomic Map