Assay Details
| Gene Symbol: Atp6v1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007508.5 | NP_031534.2 | 4 - 5 | 499 | 102 | |
| GenBank mRNA | AK140873.1 | - | 4 - 5 | 475 | 102 | |
| AK149833.1 | - | 4 - 5 | 514 | 102 | ||
| AK150742.1 | - | 4 - 5 | 485 | 102 | ||
| AK153403.1 | - | 4 - 5 | 500 | 102 | ||
| AK154869.1 | - | 4 - 5 | 500 | 102 | ||
| AK160792.1 | - | 4 - 5 | 514 | 102 | ||
| AK166857.1 | - | 4 - 5 | 489 | 102 | ||
| AK170721.1 | - | 4 - 5 | 500 | 102 | ||
| AK216026.1 | - | 1 - 2 | 152 | 102 | ||
| BC038392.1 | - | 4 - 5 | 477 | 2651677 | 102 | |
More Information
Updated Mapping Information
Set Membership: |
|

Atp6v1a
Genomic Map