Assay Details
| Gene Symbol: BC027231 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145972.4 | NP_666084.1 | 2 - 3 | 319 | 88 | |
| GenBank mRNA | AK137274.1 | - | 1 - 2 | 1012 | 88 | |
| AK144612.1 | - | 2 - 3 | 300 | 88 | ||
| AK162291.1 | - | 1 - 2 | 161 | 88 | ||
| BB651637.1 | - | 2 - 3 | 317 | 88 | ||
| BC027231.1 | - | 2 - 3 | 322 | 3585175 | 88 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK137274.1 transcripts.
Set Membership: |
|

BC027231
Genomic Map