| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1600013381.mm.1 |
NM_001159419.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600013381.mm.1 |
NM_001159419.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600013381.mm.1 |
NM_198034.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600013381.mm.1 |
NM_198034.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600013381.mm.1 |
TC1600001642.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600013382.mm.1 |
NM_001159419.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600013382.mm.1 |
NM_001159419.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600013382.mm.1 |
NM_198034.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600013382.mm.1 |
NM_198034.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600013382.mm.1 |
TC1600001642.mm.1 |
MTA TC |