Assay Details
| Gene Symbol: C330027C09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172616.2 | NP_766204.2 | 12 - 13 | 1615 | 128 | |
| GenBank mRNA | AK049356.1 | - | 12 - 13 | 1617 | 128 | |
| AK160639.1 | - | 12 - 13 | 1602 | 128 | ||
| AK173190.1 | - | 11 - 12 | 1353 | 128 | ||
| BC021874.1 | - | 1 - 2 | 810 | 128 | ||
| BC052662.1 | - | 3 - 4 | 291 | 128 | ||
| BC137719.1 | - | 12 - 13 | 1624 | 8860738 | 128 | |
More Information
Updated Mapping Information
Set Membership: |
|

C330027C09Rik
Genomic Map