Assay Details
| Gene Symbol: C330027C09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172616.2 | NP_766204.2 | 9 - 10 | 1200 | 75 | |
| GenBank mRNA | AK049356.1 | - | 9 - 10 | 1202 | 75 | |
| AK160639.1 | - | 9 - 10 | 1186 | 75 | ||
| AK173190.1 | - | 8 - 9 | 938 | 75 | ||
| BC137719.1 | - | 9 - 10 | 1209 | 8860738 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

C330027C09Rik
Genomic Map