Assay Details
| Gene Symbol: Cdip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025670.4 | NP_079946.2 | 2 - 3 | 170 | 89 | |
| GenBank mRNA | AK005157.1 | - | 2 - 3 | 156 | 89 | |
| AK017480.1 | - | 2 - 3 | 153 | 89 | ||
| AK031705.1 | - | 2 - 3 | 172 | 89 | ||
| AK035791.1 | - | 1 - 2 | 844 | 89 | ||
| AK051560.1 | - | 2 - 3 | 168 | 89 | ||
| AK149939.1 | - | 2 - 3 | 166 | 89 | ||
| AK150219.1 | - | 2 - 3 | 166 | 89 | ||
| AK166141.1 | - | 2 - 3 | 163 | 89 | ||
| BC004063.1 | - | 2 - 3 | 169 | 3590511 | 89 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK005157.1, AK017480.1, AK031705.1, AK051560.1, AK149939.1, AK150219.1, AK166141.1, BC004063.1, NM_025670.4 transcripts.
Set Membership: |
|

Cdip1
Genomic Map