Assay Details
| Gene Symbol: Alg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145362.2 | NP_663337.2 | 7 - 8 | 894 | 89 | |
| GenBank mRNA | AK145871.1 | - | 7 - 8 | 894 | 89 | |
| BC011281.1 | - | 6 - 7 | 801 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Alg1
Genomic Map