Assay Details
| Gene Symbol: Ubn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026666.3 | NP_080942.1 | 11 - 12 | 2299 | 61 | |
| GenBank mRNA | AK147571.1 | - | 11 - 12 | 2300 | 61 | |
| BC060723.1 | - | 9 - 10 | 1227 | 61 | ||
| BC098372.1 | - | 9 - 10 | 1459 | 30363389 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ubn1
Genomic Map