Assay Details
| Gene Symbol: Ubn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026666.3 | NP_080942.1 | 14 - 15 | 2622 | 133 | |
| GenBank mRNA | AK137727.1 | - | 2 - 3 | 88 | 133 | |
| AK147571.1 | - | 14 - 15 | 2623 | 133 | ||
| BC060723.1 | - | 12 - 13 | 1550 | 133 | ||
| BC098372.1 | - | 12 - 13 | 1782 | 30363389 | 133 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ubn1
Genomic Map