Assay Details
| Gene Symbol: Ubn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026666.3 | NP_080942.1 | 15 - 16 | 2661 | 114 | |
| GenBank mRNA | AK137727.1 | - | 3 - 4 | 127 | 114 | |
| AK147571.1 | - | 15 - 16 | 2662 | 114 | ||
| BC060723.1 | - | 13 - 14 | 1589 | 114 | ||
| BC098372.1 | - | 13 - 14 | 1821 | 30363389 | 114 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ubn1
Genomic Map