Assay Details
| Gene Symbol: Ubn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026666.3 | NP_080942.1 | 17 - 18 | 4137 | 94 | |
| GenBank mRNA | AK084046.1 | - | 3 - 4 | 1499 | 94 | |
| AK147571.1 | - | 17 - 18 | 4138 | 94 | ||
| AK212227.1 | - | 1 - 2 | 236 | 95 | ||
| BC098372.1 | - | 15 - 16 | 3297 | 30363389 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ubn1
Genomic Map