| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1600000405.mm.1 |
NM_026666.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600000405.mm.1 |
NM_026666.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600000405.mm.1 |
TC1600000044.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600000402.mm.1 |
NM_026666.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600000402.mm.1 |
NM_026666.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600000402.mm.1 |
TC1600000044.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600000404.mm.1 |
NM_026666.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600000404.mm.1 |
NM_026666.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600000404.mm.1 |
TC1600000044.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600000401.mm.1 |
NM_026666.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600000401.mm.1 |
NM_026666.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600000401.mm.1 |
TC1600000044.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600000406.mm.1 |
NM_026666.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600000406.mm.1 |
NM_026666.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600000406.mm.1 |
TC1600000044.mm.1 |
MTA TC |