Assay Details
| Gene Symbol: Ubn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026666.3 | NP_080942.1 | 5 - 6 | 1300 | 119 | |
| GenBank mRNA | AK019173.1 | - | 4 - 5 | 1198 | 119 | |
| AK147571.1 | - | 5 - 6 | 1301 | 119 | ||
| BC060723.1 | - | 3 - 4 | 228 | 119 | ||
| BC098372.1 | - | 3 - 4 | 460 | 30363389 | 119 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ubn1
Genomic Map