Assay Details
| Gene Symbol: Chmp2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026879.2 | NP_081155.1 | 2 - 3 | 312 | 83 | |
| GenBank mRNA | AK084111.1 | - | 2 - 3 | 314 | 83 | |
| AK180291.1 | - | 2 - 3 | 131 | 83 | ||
| AK197280.1 | - | 2 - 3 | 131 | 83 | ||
| AK201812.1 | - | 2 - 3 | 129 | 83 | ||
| AK217905.1 | - | 2 - 3 | 132 | 83 | ||
| BC042626.1 | - | 1 - 2 | 68 | 83 | ||
| BC055809.1 | - | 2 - 3 | 341 | 6414077 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Chmp2b
Genomic Map